Learn More
Medchemexpress LLC HY-44234A 5mg Medchemexpress, Fmoc-Gly-Gly-D-Phe-OtBu CAS: Purity:>98%
SDP
Catalog No. 501970066
Encompass_Preferred
Medchemexpress, HY-44234A 5mg Fmoc-Gly-Gly-D-Phe-OtBu CAS: Fmoc-Gly-Gly-D-Phe-OtBu is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Fmoc-Gly-Gly-D-Phe-OtBu is the R-isomer of Fmoc-Gly-Gly-Phe-OtBu (HY-44234). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0066 Supplier Medchemexpress LLC Supplier No. HY44234A5MG
Medchemexpress, HY-44234A 5mg Fmoc-Gly-Gly-D-Phe-OtBu CAS: Fmoc-Gly-Gly-D-Phe-OtBu is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Fmoc-Gly-Gly-D-Phe-OtBu is the R-isomer of Fmoc-Gly-Gly-Phe-OtBu (HY-44234). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.