Learn More
Medchemexpress LLC Fmoc-Gly-Gly-Gly-OH 5g
SDP
Catalog No. 5000205162
Encompass_Preferred
Fmoc-Gly-Gly-Gly-OH is a tripeptide that can be used in peptide synthesis
Catalog No. 50-002-05162 Supplier Medchemexpress LLC Supplier No. HYW0971225G
Fmoc-Gly-Gly-Gly-OH is a tripeptide that can be used in peptide synthesis