Learn More
Medchemexpress LLC Fmoc-Gly-Gly-Phe-Gly-CH2-O-CH2-Cbz | 2264011-97-2 | 97.7% | C40H41N5O9 | 100 MG
SDP
Catalog No. 5000358208
Encompass_Preferred
Fmoc-Gly-Gly-Phe-Gly-CH2-O-CH2-Cbz is a PROTAC linker and a maleimide-GGFG peptide linker. It can be utilized in the synthesis of Deruxtecan. This product is intended for research use only.
  • PROTAC linker
  • Maleimide-GGFG peptide linker
  • Can be used in the synthesis of Deruxtecan
  • Intended for research use only
Catalog No. 50-003-58208 Supplier Medchemexpress LLC Supplier No. HY45669100MG
Fmoc-Gly-Gly-Phe-Gly-CH2-O-CH2-Cbz is a PROTAC linker and a maleimide-GGFG peptide linker. It can be utilized in the synthesis of Deruxtecan. This product is intended for research use only.
  • PROTAC linker
  • Maleimide-GGFG peptide linker
  • Can be used in the synthesis of Deruxtecan
  • Intended for research use only