Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CO-CH3 | 2866301-96-2 | 98.7% | C33H35N5O8 | 25 MG
SDP

Catalog No. 5000358927
Encompass_Preferred

Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CO-CH3 (compound DC-13-C) is an intermediate used in the synthesis of Exatecan derivatives. This solid compound has a molecular weight of 629.66 and typically appears white to off-white. For optimal preservation, store powder at -20°C for up to 3 years. If dissolved, store in solvent at -80°C for 6 months or -20°C for 1 month. It exhibits a solubility of 100 mg/mL (158.82 mM) in DMSO.

  • Molecular weight: 629.66
  • Appearance: Solid
  • Color: White to off-white
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months or -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL (158.82 mM)
  • Biological activity: Drug intermediate

Catalog No. 50-003-58927 Supplier Medchemexpress LLC Supplier No. 25MGHY49412
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CO-CH3 (compound DC-13-C) is an intermediate used in the synthesis of Exatecan derivatives. This solid compound has a molecular weight of 629.66 and typically appears white to off-white. For optimal preservation, store powder at -20°C for up to 3 years. If dissolved, store in solvent at -80°C for 6 months or -20°C for 1 month. It exhibits a solubility of 100 mg/mL (158.82 mM) in DMSO.

  • Molecular weight: 629.66
  • Appearance: Solid
  • Color: White to off-white
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months or -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL (158.82 mM)
  • Biological activity: Drug intermediate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.