Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CO-CH3 | 2866301-96-2 | 98.7% | C33H35N5O8 | 50 MG
SDP

Catalog No. 5000357716
Encompass_Preferred

Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CO-CH3, also known as compound DC-13-C, is an intermediate used in the synthesis of Exatecan derivatives.

  • Purity of 98.68%
  • Appears as a white to off-white solid
  • Soluble in DMSO (100 mg/mL)
  • Powder stable for up to 3 years at -20°C
  • Solution stable for up to 6 months at -80°C or 1 month at -20°C

Catalog No. 50-003-57716 Supplier Medchemexpress LLC Supplier No. HY4941250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CO-CH3, also known as compound DC-13-C, is an intermediate used in the synthesis of Exatecan derivatives.

  • Purity of 98.68%
  • Appears as a white to off-white solid
  • Soluble in DMSO (100 mg/mL)
  • Powder stable for up to 3 years at -20°C
  • Solution stable for up to 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.