Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Gly-NH-CH2-O-cyclopropane-CH2COOH | 2414254-47-8 | 96.0% | 424.45 g/mol | C23H24N2O6 | 5 MG
SDP

Catalog No. 5000281610
Encompass_Preferred

Fmoc-Gly-NH-CH2-O-Cyclopropane-CH2COOH is an Fmoc-protected glycine derivative used as a cleavable linker intermediate in the synthesis of antibody-drug conjugates and related bioconjugation applications. Supplied as a solid with supporting analytical data, it is intended for research use only and includes documentation for quality and handling.

  • Contains a cyclopropane spacer for linker design.
  • Fmoc-protected glycine suitable for solid-phase peptide synthesis.
  • Supplied as a solid, white to off-white in appearance.
  • Purity approximately 96.0%.
  • Molecular weight 424.45 g/mol; formula C23H24N2O6.
  • Store at 4°C under nitrogen; in solvent keep at -80°C for long-term storage.
  • Includes COA, SDS, HNMR, and RP-HPLC documentation.

Catalog No. 50-002-81610 Supplier Medchemexpress LLC Supplier No. HYW2505415MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Gly-NH-CH2-O-Cyclopropane-CH2COOH is an Fmoc-protected glycine derivative used as a cleavable linker intermediate in the synthesis of antibody-drug conjugates and related bioconjugation applications. Supplied as a solid with supporting analytical data, it is intended for research use only and includes documentation for quality and handling.

  • Contains a cyclopropane spacer for linker design.
  • Fmoc-protected glycine suitable for solid-phase peptide synthesis.
  • Supplied as a solid, white to off-white in appearance.
  • Purity approximately 96.0%.
  • Molecular weight 424.45 g/mol; formula C23H24N2O6.
  • Store at 4°C under nitrogen; in solvent keep at -80°C for long-term storage.
  • Includes COA, SDS, HNMR, and RP-HPLC documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.