Learn More
Apexbio Technology LLC Fmoc-Gly-OH 29022-11-5 250g
SDP
Catalog No. 5000088661
Encompass_Preferred
Fmoc-Gly-OH (29022-11-5) is an Fmoc-protected glycine derivative commonly used in solid-phase peptide synthesis (SPPS) Structurally it features the fluorenylmethyloxycarbonyl (Fmoc) group shielding the amino terminus which can be selectively removed under mild alkaline conditions without compromising peptide integrity This selective deprotection allows for precise stepwise assembly of peptide chains Based on these properties Fmoc-Gly-OH is widely applied in biomedical research including pharmaceutical design peptide synthesis structural characterization and studies on protein function and interactions
Catalog No. 50-000-88661 Supplier Apexbio Technology LLC Supplier No. A7901250000
Fmoc-Gly-OH (29022-11-5) is an Fmoc-protected glycine derivative commonly used in solid-phase peptide synthesis (SPPS) Structurally it features the fluorenylmethyloxycarbonyl (Fmoc) group shielding the amino terminus which can be selectively removed under mild alkaline conditions without compromising peptide integrity This selective deprotection allows for precise stepwise assembly of peptide chains Based on these properties Fmoc-Gly-OH is widely applied in biomedical research including pharmaceutical design peptide synthesis structural characterization and studies on protein function and interactions