Learn More
Bachem Fmoc-Gly-2-methoxy-4-alkoxybenzyl alcohol resin
SDP
Supplier:  Bachem 4012730.0005
Encompass_Preferred
Fmoc-Gly-SASRINTM resin (200-400 mesh, 0.5-0.8 mmol/g) , Fmoc-Gly-2-methoxy-4-alkoxybenzyl alcohol resin, , n/a, Mr n/a, 5g.
Catalog No. 50-258-895