Learn More
Apexbio Technology LLC Fmoc-Gly-Wang resin 25g
SDP
Catalog No. 5000061603
Encompass_Preferred
Fmoc-Gly-Wang resin is a solid-phase support featuring glycine residues bound onto a polymer matrix with amino groups protected by a 9-fluorenylmethoxycarbonyl (Fmoc) moiety In solid-phase peptide synthesis (SPPS) this resin facilitates the stepwise elongation of peptide chains through iterative cycles involving Fmoc-deprotection and subsequent amino acid coupling reactions After peptide assembly cleavage under acidic conditions releases the synthesized peptide from the resin support Given its defined chemical structure and stability during synthesis this resin is widely utilized in biomedical research fields including studies of protein structure-function relationships bioactive peptide design and therapeutic peptide development
Catalog No. 50-000-61603 Supplier Apexbio Technology LLC Supplier No. A683925000
Fmoc-Gly-Wang resin is a solid-phase support featuring glycine residues bound onto a polymer matrix with amino groups protected by a 9-fluorenylmethoxycarbonyl (Fmoc) moiety In solid-phase peptide synthesis (SPPS) this resin facilitates the stepwise elongation of peptide chains through iterative cycles involving Fmoc-deprotection and subsequent amino acid coupling reactions After peptide assembly cleavage under acidic conditions releases the synthesized peptide from the resin support Given its defined chemical structure and stability during synthesis this resin is widely utilized in biomedical research fields including studies of protein structure-function relationships bioactive peptide design and therapeutic peptide development