Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Gly3-Val-Cit-PAB | 2647914-09-6 | 95.0% | 772.85 g/mol | C39H48N8O9 | 5 MG
SDP

Catalog No. 5000203866
Encompass_Preferred

Fmoc-Gly3-Val-Cit-PAB is a Fmoc-protected, cleavable peptide-based ADC linker used for the synthesis of antibody-drug conjugates (ADCs). It contains a Val-Cit cleavable dipeptide and a para-aminobenzyl (PAB) self-immolative spacer and is supplied at 95.0% purity.

  • Cleavable Val-Cit peptide linker.
  • Fmoc-protected for solid-phase synthesis compatibility.
  • Contains para-aminobenzyl (PAB) self-immolative spacer.
  • Molecular weight 772.85 g/mol.
  • Molecular formula C39H48N8O9.
  • Available in small-scale research quantities (e.g., 5 mg).

Catalog No. 50-002-03866 Supplier Medchemexpress LLC Supplier No. HY1361065MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Gly3-Val-Cit-PAB is a Fmoc-protected, cleavable peptide-based ADC linker used for the synthesis of antibody-drug conjugates (ADCs). It contains a Val-Cit cleavable dipeptide and a para-aminobenzyl (PAB) self-immolative spacer and is supplied at 95.0% purity.

  • Cleavable Val-Cit peptide linker.
  • Fmoc-protected for solid-phase synthesis compatibility.
  • Contains para-aminobenzyl (PAB) self-immolative spacer.
  • Molecular weight 772.85 g/mol.
  • Molecular formula C39H48N8O9.
  • Available in small-scale research quantities (e.g., 5 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.