Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Gly3-Val-Cit-PAB | 2647914-09-6 | MFCD30179684 | 95.0% | 772.85 g/mol | C39H48N8O9 | 10 MG
SDP

Catalog No. 5000203905
Encompass_Preferred

Fmoc-Gly3-Val-Cit-PAB is a peptide-based, protease-cleavable linker used for assembling antibody-drug conjugates. It is supplied as a purified solid with defined purity and formulation guidance for conjugation and in vivo formulation.

  • Cleavable linker designed for protease-mediated payload release.
  • Fmoc-protected peptide structure for solid-phase or solution conjugation workflows.
  • High purity suitable for research conjugation (reported 95.0%).
  • Supplied as a white to off-white solid, facilitating handling and storage.
  • Soluble in DMSO at high concentration, with manufacturer-recommended formulation examples for in vivo use.
  • Storage recommendations include refrigerated and low-temperature solvent conditions to preserve stability.
  • Available in small pack sizes suitable for research-scale conjugation work.

Catalog No. 50-002-03905 Supplier Medchemexpress LLC Supplier No. HY13610610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Gly3-Val-Cit-PAB is a peptide-based, protease-cleavable linker used for assembling antibody-drug conjugates. It is supplied as a purified solid with defined purity and formulation guidance for conjugation and in vivo formulation.

  • Cleavable linker designed for protease-mediated payload release.
  • Fmoc-protected peptide structure for solid-phase or solution conjugation workflows.
  • High purity suitable for research conjugation (reported 95.0%).
  • Supplied as a white to off-white solid, facilitating handling and storage.
  • Soluble in DMSO at high concentration, with manufacturer-recommended formulation examples for in vivo use.
  • Storage recommendations include refrigerated and low-temperature solvent conditions to preserve stability.
  • Available in small pack sizes suitable for research-scale conjugation work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.