Learn More
Medchemexpress LLC Fmoc-glycine-2,2-d2 | 284665-11-8 | MFCD01075436 | 99.5% | 299.32 | C17H13D2NO4 | 50mg
SDP
Catalog No. 5000267457
Encompass_Preferred
Fmoc-Gly-OH-2 2-d2 (Fmoc-glycine-2 2-d2) is the deuterium labeled Fmoc-Gly-OH (HY-Y1250) Fmoc-Gly-OH (Fmoc glycine) is a Fmoc-protected glycine derivative can be used for the synthesis of compounds[1]
Catalog No. 50-002-67457 Supplier Medchemexpress LLC Supplier No. HYY1250S250MG
Fmoc-Gly-OH-2 2-d2 (Fmoc-glycine-2 2-d2) is the deuterium labeled Fmoc-Gly-OH (HY-Y1250) Fmoc-Gly-OH (Fmoc glycine) is a Fmoc-protected glycine derivative can be used for the synthesis of compounds[1]