Learn More
Chem-Impex International, Inc. Fmoc-glycine-2-chlorotrityl resin (0.3 - 1.0 meq/g, 200 - 400 mesh) | MFCD00801575 | 1G
Supplier:  Chem-Impex International, Inc. 042321G
Encompass_Preferred
Fmoc-glycine-2-chlorotrityl resin (0.3 - 1.0 meq/g, 200 - 400 mesh), MFCD00801575, 1G
Catalog No. 50-499-411