Learn More
Bachem Fmoc-His(1-Boc)-OH
SDP
Catalog No. 50258586
Encompass_Preferred
Fmoc-His(1-Boc)-OH, 81379-52-4, C26H27N3O6, Mr 477.52, 25g. Fmoc-His(t-Boc)-OH. Fmoc-His(Boc)-OH shows a lower tendency to racemize during coupling than the Trt analog. The derivative has been used in the SPPS of oligohistidines and peptide-oligonucleotide conjugates.
Catalog No. 50-258-586 Supplier Bachem Supplier No. 4010990.0025
Add to Cart
Add to Cart
Fmoc-His(1-Boc)-OH, 81379-52-4, C26H27N3O6, Mr 477.52, 25g. Fmoc-His(t-Boc)-OH. Fmoc-His(Boc)-OH shows a lower tendency to racemize during coupling than the Trt analog. The derivative has been used in the SPPS of oligohistidines and peptide-oligonucleotide conjugates.