Learn More
Bachem Fmoc-His(1-Trt)-OH
SDP
Supplier:  Bachem 4011444.0100
Encompass_Preferred
Fmoc-His(1-Trt)-OH, 109425-51-6, C40H33N3O4, Mr 619.72, 100g. Fmoc-His(t-Trt)-OH. As to minimize racemization, Fmoc-His(Trt)-OH is best coupled with DEPBT.
Catalog No. 50-258-554