Learn More
Bachem Fmoc-His(1-Trt)-OH
SDP
Catalog No. 50258553
Encompass_Preferred
Fmoc-His(1-Trt)-OH, 109425-51-6, C40H33N3O4, Mr 619.72, 25g. Fmoc-His(t-Trt)-OH. As to minimize racemization, Fmoc-His(Trt)-OH is best coupled with DEPBT.
Catalog No. 50-258-553 Supplier Bachem Supplier No. 4011444.0025
Add to Cart
Add to Cart
Fmoc-His(1-Trt)-OH, 109425-51-6, C40H33N3O4, Mr 619.72, 25g. Fmoc-His(t-Trt)-OH. As to minimize racemization, Fmoc-His(Trt)-OH is best coupled with DEPBT.