Learn More
Apexbio Technology LLC Fmoc-β-homo-Arg(Pbf)-OH 401915-53-5 5g
SDP
Catalog No. 5000093460
Encompass_Preferred
Fmoc-beta-homo-Arg Pbf -OH (CAS 401915-53-5) is designed as an Fmoc-protected -amino acid derivative containing an arginine side chain analog with a Pbf-protected guanidino group It is intended for incorporation into peptides via solid-phase peptide synthesis (SPPS) enabling the systematic modification of peptide scaffolds for structural studies Based on these properties Fmoc-beta-homo-Arg Pbf -OH supports research in the preparation of -amino acid peptide analogs with applications in exploring protein-protein interactions biomolecule recognition stability biological activity and development of enzyme inhibitors or peptide analogs for mechanistic studies at the molecular and cellular levels
Catalog No. 50-000-93460 Supplier Apexbio Technology LLC Supplier No. A67055000
Fmoc-beta-homo-Arg Pbf -OH (CAS 401915-53-5) is designed as an Fmoc-protected -amino acid derivative containing an arginine side chain analog with a Pbf-protected guanidino group It is intended for incorporation into peptides via solid-phase peptide synthesis (SPPS) enabling the systematic modification of peptide scaffolds for structural studies Based on these properties Fmoc-beta-homo-Arg Pbf -OH supports research in the preparation of -amino acid peptide analogs with applications in exploring protein-protein interactions biomolecule recognition stability biological activity and development of enzyme inhibitors or peptide analogs for mechanistic studies at the molecular and cellular levels