Learn More
Bachem Fmoc-Homoleu-OH
SDP
Supplier:  Bachem 4039689.0001
Encompass_Preferred
Fmoc-Homoleu-OH, 180414-94-2, C22H25NO4, Mr 367.44, 1g. Fmoc-Hle-OH. Hle has been incorporated as a replacement of Met35 in Ab 1-40 and 1-42 analogs produced for studying the role of the Met residue in controlling Ab oligomerization.
Catalog No. 50-312-952