Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Hse Trt -OH, Advanced ChemTech

Catalog No. 01340028
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-028 1 g
01-340-027 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-340-028 Supplier Advanced Chem Tech Supplier No. FS24431G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 111061-55-3
Molecular Formula C38H33NO5
Molecular Weight (g/mol) 583.68
MDL Number MFCD00270544
InChI Key QUTREFXHXPNEJN-DHUJRADRSA-N
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(triphenylmethoxy)butanoic acid
SMILES OC(=O)[C@H](CCOC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White/Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-Hse Trt -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.