Learn More
MilliporeSigma FMOC-HYP(TBU)-OH
Catalog No. 502301741
Encompass
Fmoc-Hyp(tBu)-OH; > /=98% by TLC and HPLC; Solid; EMD; Novabiochem; 1g; N-alpha-Fmoc-O-tert-butyl-L-trans-4-hydroxyproline; F.W. 409.5; C24H27NO5; CAS: 122996-47-8
Catalog No. 50-230-1741 Supplier MilliporeSigma Supplier No. 8520360001
Fmoc-Hyp(tBu)-OH; > /=98% by TLC and HPLC; Solid; EMD; Novabiochem; 1g; N-alpha-Fmoc-O-tert-butyl-L-trans-4-hydroxyproline; F.W. 409.5; C24H27NO5; CAS: 122996-47-8