Learn More
Apexbio Technology LLC Fmoc-Ile-OPfp 86060-89-1 25g
SDP
Catalog No. 5000090583
Encompass_Preferred
Fmoc-Ile-OPfp is an amino acid derivative containing an Fmoc-protected isoleucine residue and a pentafluorophenyl ester (OPfp) functional group widely utilized in the solid-phase peptide synthesis (SPPS) process It facilitates peptide bond formation during coupling reactions via its activated pentafluorophenyl ester moiety reacting with free amine groups on immobilized peptide chains or amino acid residues The Fmoc protecting group allows for sequential amino acid addition by selective cleavage under mild basic conditions Fmoc-Ile-OPfp supports the controlled synthesis of peptides for applications in medicinal chemistry research functional peptide fragment studies protein structure analysis and biochemical investigations of peptide-mediated biological activities
Catalog No. 50-000-90583 Supplier Apexbio Technology LLC Supplier No. A790925000
Fmoc-Ile-OPfp is an amino acid derivative containing an Fmoc-protected isoleucine residue and a pentafluorophenyl ester (OPfp) functional group widely utilized in the solid-phase peptide synthesis (SPPS) process It facilitates peptide bond formation during coupling reactions via its activated pentafluorophenyl ester moiety reacting with free amine groups on immobilized peptide chains or amino acid residues The Fmoc protecting group allows for sequential amino acid addition by selective cleavage under mild basic conditions Fmoc-Ile-OPfp supports the controlled synthesis of peptides for applications in medicinal chemistry research functional peptide fragment studies protein structure analysis and biochemical investigations of peptide-mediated biological activities