Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMOC isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog No. AC438660010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438660010 1 g
AC438660050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438660010 Supplier Thermo Scientific Chemicals Supplier No. 438660010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 199915-38-3 | Molecular Formula: C16H11NO2S | Molecular Weight: 281.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199915-38-3
Molecular Formula C16H11NO2S
Molecular Weight (g/mol) 281.34
InChI Key DHMYULZVFHHEHE-UHFFFAOYSA-N
Synonym fmoc isothiocyanate, fmoc-isothiocyanate, 9-fluorenylmethoxycarbonyl isothiocyanate, 9h-fluoren-9-ylmethyl n-sulfanylidenemethylidene carbamate, 9h-fluoren-9-ylmethyl n-carbothioylcarbamate, n-fmoc-isothiocyanate, 9-fluorenylmethyl isothiocyanatoformate, 9-fluorenylmethoxycarbonylisothiocyanate, fmoc isothiocyanate chn, fluoren-9-ylmethyloxycarbonyl isothiocyanate
PubChem CID 4154759
IUPAC Name 9H-fluoren-9-ylmethyl N-(sulfanylidenemethylidene)carbamate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N=C=S

Specifications

Melting Point 44.0°C to 45.0°C
Color Yellow to Brown
Elemental Analysis 65.6 to 71.0 % (C) 4.8 to 5.2 % (N)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 281.34
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material FMOC isothiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.