Learn More
AnaSpec FMOC-L-(2, 6-DI-ME)TYR-OH
Catalog No. 50352439
Encompass
Fmoc-L-(2,6-di-Me)Tyr-OH, ; MW 431.49; (1 g) 2,6-dimethyl-tyrosine(Dmt) is chi-constrained a -amino acid, and its incorporation in strategic positions of peptides(local side-chain constraines) has allowed for a substantial reduction of the corresponding side-chain conformers.
Catalog No. 50352439 Supplier AnaSpec Supplier No. AS637291
Fmoc-L-(2,6-di-Me)Tyr-OH, ; MW 431.49; (1 g) 2,6-dimethyl-tyrosine(Dmt) is chi-constrained a -amino acid, and its incorporation in strategic positions of peptides(local side-chain constraines) has allowed for a substantial reduction of the corresponding side-chain conformers.