Learn More
BLD PHARMATECH CO LIMITED Fmoc-L-Arg(Pbf)-Wang Resin, 5g, Purity: 0.25-0.8mmol/g
Catalog No. NC3965224
Encompass_Preferred
Fmoc-L-Arg(Pbf)-Wang Resin, 5g, Purity: 0.25-0.8mmol/g
Catalog No. NC3965224 Supplier BLD PHARMATECH CO LIMITED Supplier No. BD3024235G
Fmoc-L-Arg(Pbf)-Wang Resin, 5g, Purity: 0.25-0.8mmol/g