Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-L-Arg(Pmc)-OH | 50 g | CAS 119831-72-0 | MDL MFCD00065618

Catalog No. 502357248
Encompass

Fmoc-L-Arg(Pmc)-OH is a Amino Acid reagent (Subcategory: Arg) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 119831-72-0
- MDL: MFCD00065618
- InChIKey: QTWZCODKTSUZJN-LJAQVGFWSA-N
- Molecular Weight: 662.802
- Molecular Formula: C35H42N4O7S
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29359090
- Country Of Origin: China
- IUPAC: N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-Nw-((2,2,5,7,8-pentamethylchroman-6-yl)sulfonyl)-L-arginine
- SMILES: CC1=C2C(CCC(O2)(C)C)=C(C)C(S(=O)(NC(NCCC[C@@H](C(O)=O)NC(OCC3C4=CC=CC=C4C5=CC=CC=C53)=O)=N)=O)=C1C

Catalog No. 50-235-7248 Supplier STA Pharmaceutical Supplier No. STAAA1205450GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-L-Arg(Pmc)-OH is a Amino Acid reagent (Subcategory: Arg) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 119831-72-0
- MDL: MFCD00065618
- InChIKey: QTWZCODKTSUZJN-LJAQVGFWSA-N
- Molecular Weight: 662.802
- Molecular Formula: C35H42N4O7S
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29359090
- Country Of Origin: China
- IUPAC: N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-Nw-((2,2,5,7,8-pentamethylchroman-6-yl)sulfonyl)-L-arginine
- SMILES: CC1=C2C(CCC(O2)(C)C)=C(C)C(S(=O)(NC(NCCC[C@@H](C(O)=O)NC(OCC3C4=CC=CC=C4C5=CC=CC=C53)=O)=N)=O)=C1C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.