Learn More
Medchemexpress LLC Fmoc-L-Asn(beta-D-GlcNAc(Ac)3)-OH | 131287-39-3 | 95.03% | C33H37N3O13 | 50 MG
SDP
Catalog No. 5000356776
Encompass_Preferred
Fmoc-L-Asn(beta-D-GlcNAc(Ac)3)-OH is an amino acid derivative used in the synthesis of silicon-fluoride acceptor (SiFA) derivatized octreotate derivatives. These SiFA-octreotate analogues serve as tumor imaging agents and are valuable tools for research in positron emission tomography (PET).
  • Amino acid derivative
  • Used in the synthesis of silicon-fluoride acceptor (SiFA) derivatized octreotate derivatives
  • Serves as a tumor imaging agent
  • Useful tool for positron emission tomography (PET) research
Catalog No. 50-003-56776 Supplier Medchemexpress LLC Supplier No. HY14146450MG
Fmoc-L-Asn(beta-D-GlcNAc(Ac)3)-OH is an amino acid derivative used in the synthesis of silicon-fluoride acceptor (SiFA) derivatized octreotate derivatives. These SiFA-octreotate analogues serve as tumor imaging agents and are valuable tools for research in positron emission tomography (PET).
  • Amino acid derivative
  • Used in the synthesis of silicon-fluoride acceptor (SiFA) derivatized octreotate derivatives
  • Serves as a tumor imaging agent
  • Useful tool for positron emission tomography (PET) research