Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-L-Asn(beta-D-GlcNAc(Ac)3)-OH | 131287-39-3 | C33H37N3O13 | 1 ML
SDP

Catalog No. 5000357063
Encompass_Preferred

Fmoc-L-Asn(beta-D-GlcNAc(Ac)3)-OH is a compound used in the synthesis of silicon-fluoride acceptor (SIFA) derivatized octreotate derivatives. These SiFA-octreotate analogues serve as tumor imaging agents, providing a useful tool for research in positron emission tomography (PET).

  • CAS number: 131287-39-3
  • Molecular formula: C33H37N3O13
  • Molecular weight: 683.66
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 125 mg/mL (182.84 mM)

Catalog No. 50-003-57063 Supplier Medchemexpress LLC Supplier No. HY14146410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-L-Asn(beta-D-GlcNAc(Ac)3)-OH is a compound used in the synthesis of silicon-fluoride acceptor (SIFA) derivatized octreotate derivatives. These SiFA-octreotate analogues serve as tumor imaging agents, providing a useful tool for research in positron emission tomography (PET).

  • CAS number: 131287-39-3
  • Molecular formula: C33H37N3O13
  • Molecular weight: 683.66
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 125 mg/mL (182.84 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.