Learn More
BLD PHARMATECH CO LIMITED Fmoc-L-Asn(Trt)-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g
Catalog No. NC3965355
Encompass_Preferred
Fmoc-L-Asn(Trt)-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g
Catalog No. NC3965355 Supplier BLD PHARMATECH CO LIMITED Supplier No. BD3024255G
Add to Cart
Add to Cart
Fmoc-L-Asn(Trt)-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g