Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-L-Asp-OMe | 25 g | CAS 145038-52-4 | MDL MFCD03701473

Catalog No. 502350568
Encompass

Fmoc-L-Asp-OMe is a Amino Acid reagent (Subcategory: Asp) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 145038-52-4
- MDL: MFCD03701473
- InChIKey: UEUZUMRMWJUEMK-KRWDZBQOSA-N
- Molecular Weight: 369.373
- Molecular Formula: C20H19NO6
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxy-4-oxobutanoic acid
- SMILES: COC([C@H](CC(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)=O

Catalog No. 50-235-0568 Supplier STA Pharmaceutical Supplier No. STAAA1063825GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-L-Asp-OMe is a Amino Acid reagent (Subcategory: Asp) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 145038-52-4
- MDL: MFCD03701473
- InChIKey: UEUZUMRMWJUEMK-KRWDZBQOSA-N
- Molecular Weight: 369.373
- Molecular Formula: C20H19NO6
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxy-4-oxobutanoic acid
- SMILES: COC([C@H](CC(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.