Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-L-β-homoarginine(Pmc) | 50 g | CAS 700377-76-0 | MDL MFCD03788172

Catalog No. 502352996
Encompass

Fmoc-L-β-homoarginine(Pmc) is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 700377-76-0
- MDL: MFCD03788172
- InChIKey: MGKMEQWQKICWRD-DEOSSOPVSA-N
- Molecular Weight: 676.829
- Molecular Formula: C36H44N4O7S
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29359090
- Country Of Origin: China
- IUPAC: (S)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-(3-((2,2,5,7,8-pentamethylchroman-6-yl)sulfonyl)guanidino)hexanoic acid
- SMILES: CC1=C2OC(C)(C)CCC2=C(C)C(=C1C)S(=O)(=O)NC(=N)NCCC[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Catalog No. 50-235-2996 Supplier STA Pharmaceutical Supplier No. STAAA1007150GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-L-β-homoarginine(Pmc) is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 700377-76-0
- MDL: MFCD03788172
- InChIKey: MGKMEQWQKICWRD-DEOSSOPVSA-N
- Molecular Weight: 676.829
- Molecular Formula: C36H44N4O7S
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29359090
- Country Of Origin: China
- IUPAC: (S)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-(3-((2,2,5,7,8-pentamethylchroman-6-yl)sulfonyl)guanidino)hexanoic acid
- SMILES: CC1=C2OC(C)(C)CCC2=C(C)C(=C1C)S(=O)(=O)NC(=N)NCCC[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.