Learn More
Medchemexpress LLC Fmoc-L-citrulline | 133174-15-9 | 397.42 | 25 G
SDP
Catalog No. 5000449819
Encompass_Preferred
Fmoc-L-Citrulline is an amino acid derivative featuring an Fmoc protecting group. It can be used to synthesize a degradable antibody-drug conjugate (ADC) linker, specifically forming a valine-citrulline (Val-Cit) motif. This linker connects an antibody to an ADC cytotoxin.
  • Amino acid derivative with Fmoc protecting group
  • Used for synthesis of degradable ADC linkers
  • Forms valine-citrulline (Val-Cit) motif
  • Stable in solid form: Store at room temperature for 3 years
  • Stable in solvent: -80°C for 2 years, -20°C for 1 year
Catalog No. 50-004-49819 Supplier Medchemexpress LLC Supplier No. 25GHYW008064
Fmoc-L-Citrulline is an amino acid derivative featuring an Fmoc protecting group. It can be used to synthesize a degradable antibody-drug conjugate (ADC) linker, specifically forming a valine-citrulline (Val-Cit) motif. This linker connects an antibody to an ADC cytotoxin.
  • Amino acid derivative with Fmoc protecting group
  • Used for synthesis of degradable ADC linkers
  • Forms valine-citrulline (Val-Cit) motif
  • Stable in solid form: Store at room temperature for 3 years
  • Stable in solvent: -80°C for 2 years, -20°C for 1 year