Learn More
Cayman Chemical Fmoc-L-Cys(Mmt)-OH, Purity: ≥98%, CAS: 177582-21-7, 50 g
SDP
Catalog No. 502945007
Encompass_Preferred
An amino acid-containing building block; has been used in the solid-phase synthesis of S-palmitoylated peptide antigens for use in macrophage uptake assays and stapled peptide dual agonists of GLP-1R and CCK2 receptors
Catalog No. 50-294-5007 Supplier Cayman Chemical Supplier No. 4217750 G
An amino acid-containing building block; has been used in the solid-phase synthesis of S-palmitoylated peptide antigens for use in macrophage uptake assays and stapled peptide dual agonists of GLP-1R and CCK2 receptors