Learn More
Medchemexpress LLC Fmoc-L-cysteic acid 50g
SDP
Catalog No. 5000205161
Encompass_Preferred
Fmoc-L-cysteic acid is an amino acid derivative with an Fmoc protecting group which can be used to synthesize triarylsulfonium compounds for radiolabeling of peptides[1]
Catalog No. 50-002-05161 Supplier Medchemexpress LLC Supplier No. HYW09705450G
Fmoc-L-cysteic acid is an amino acid derivative with an Fmoc protecting group which can be used to synthesize triarylsulfonium compounds for radiolabeling of peptides[1]