Learn More
BLD PHARMATECH CO LIMITED Fmoc-L-Gln-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g
Catalog No. NC3965283
Encompass_Preferred
Fmoc-L-Gln-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g
Catalog No. NC3965283 Supplier BLD PHARMATECH CO LIMITED Supplier No. BD011517885G
Fmoc-L-Gln-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g