Learn More
eMolecules​ Fmoc-L-Lys(Alloc)-OH | Combi-Blocks, Inc. | 146982-27-6 | MFCD00190872 | 452.507 | C25H28N2O6 | 98.000 | OC(=O)[C@H](CCCCNC(=O)OCC=C)NC(=O)OCC1c2ccccc2-c2ccccc12 | 500g | 603131051
Catalog No. NC2231566 Shop All eMolecules Inc. Products
Encompass_Preferred
Fmoc-L-Lys(Alloc)-OH | Combi-Blocks, Inc. | 146982-27-6 | MFCD00190872 | 452.507 | C25H28N2O6 | 98.000 | OC(=O)[C@H](CCCCNC(=O)OCC=C)NC(=O)OCC1c2ccccc2-c2ccccc12 | 500g | 603131051
Catalog No. NC2231566 Supplier eMolecules​ Supplier No. 603131051
Fmoc-L-Lys(Alloc)-OH | Combi-Blocks, Inc. | 146982-27-6 | MFCD00190872 | 452.507 | C25H28N2O6 | 98.000 | OC(=O)[C@H](CCCCNC(=O)OCC=C)NC(=O)OCC1c2ccccc2-c2ccccc12 | 500g | 603131051