Learn More
BLD PHARMATECH CO LIMITED Fmoc-L-Lys(Mtt)-Wang Resin, 5g, Purity: 97% 100~200mesh,1%DVB,0.3-0.8mmol/g
Catalog No. NC3965410
Encompass_Preferred
Fmoc-L-Lys(Mtt)-Wang Resin, 5g, Purity: 97% 100~200mesh,1%DVB,0.3-0.8mmol/g
Catalog No. NC3965410 Supplier BLD PHARMATECH CO LIMITED Supplier No. BD6290835G
Fmoc-L-Lys(Mtt)-Wang Resin, 5g, Purity: 97% 100~200mesh,1%DVB,0.3-0.8mmol/g