Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-L-Lys[Oct-(OtBu)-Glu-(OtBu)-AEEA-AEEA]-OH | 1662688-20-1 | MFCD34563738 | 99.7% | 1196.51 g/mol | C64H101N5O16 | 1 ML
SDP

Catalog No. 5000284842
Encompass_Preferred

Fmoc-L-Lys[Oct-(OtBu)-Glu-(OtBu)-AEEA-AEEA]-OH is an Fmoc-protected lysine derivative used as a building block in peptide synthesis, suitable for incorporation into GLP-1 analogues. It is supplied as a 10 mM solution in DMSO (1 mL) and as solids in multiple pack sizes for research use only.

  • Fmoc-protected lysine derivative for peptide synthesis.
  • Supplied as a 10 mM solution in DMSO (1 mL) and as solids in multiple pack sizes.
  • High HPLC purity (99.7%).
  • Molecular weight 1196.51 g/mol.
  • CAS number 1662688-20-1.
  • Recommended solvent DMSO; use newly opened solvent and ultrasonic agitation if required for solubility.

Catalog No. 50-002-84842 Supplier Medchemexpress LLC Supplier No. HYW61515310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-L-Lys[Oct-(OtBu)-Glu-(OtBu)-AEEA-AEEA]-OH is an Fmoc-protected lysine derivative used as a building block in peptide synthesis, suitable for incorporation into GLP-1 analogues. It is supplied as a 10 mM solution in DMSO (1 mL) and as solids in multiple pack sizes for research use only.

  • Fmoc-protected lysine derivative for peptide synthesis.
  • Supplied as a 10 mM solution in DMSO (1 mL) and as solids in multiple pack sizes.
  • High HPLC purity (99.7%).
  • Molecular weight 1196.51 g/mol.
  • CAS number 1662688-20-1.
  • Recommended solvent DMSO; use newly opened solvent and ultrasonic agitation if required for solubility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.