Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-L-N-Me-Cha-OH | 1 g | CAS 148983-03-3 | MDL MFCD00156555

Catalog No. 502359910
Encompass

Fmoc-L-N-Me-Cha-OH is a Amino Acid reagent (Subcategory: N-Me AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 148983-03-3
- MDL: MFCD00156555
- InChIKey: SZQWVTJZNYDCRV-QHCPKHFHSA-N
- Molecular Weight: 407.51
- Molecular Formula: C25H29NO4
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-cyclohexylpropanoic acid
- SMILES: CN(C(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)[C@H](C(O)=O)CC4CCCCC4

Catalog No. 50-235-9910 Supplier STA Pharmaceutical Supplier No. STAAA106521GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-L-N-Me-Cha-OH is a Amino Acid reagent (Subcategory: N-Me AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 148983-03-3
- MDL: MFCD00156555
- InChIKey: SZQWVTJZNYDCRV-QHCPKHFHSA-N
- Molecular Weight: 407.51
- Molecular Formula: C25H29NO4
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-cyclohexylpropanoic acid
- SMILES: CN(C(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)[C@H](C(O)=O)CC4CCCCC4

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.