Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-L-octahydroindole-2-carboxylic acid | 130309-37-4 | MFCD09750487 | 99.9% | 391.46 g/mol | C24H25NO4 | 5 G
SDP

Catalog No. 5000209167
Encompass_Preferred

Fmoc-Oic-OH is an Fmoc-protected amino acid derivative used as a building block in solid-phase peptide synthesis and for preparing peptide mimetics. It is supplied with high purity and supplied under recommended storage conditions.

  • Fmoc-protected amino acid for peptide synthesis.
  • High purity suitable for research use (≈99.9%).
  • Molecular weight 391.46 g/mol and formula C24H25NO4.
  • Available as a 5 g package.
  • Store at room temperature as a solid; in solvent store at -80°C or -20°C per guidelines.

Catalog No. 50-002-09167 Supplier Medchemexpress LLC Supplier No. HYW0419895G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Oic-OH is an Fmoc-protected amino acid derivative used as a building block in solid-phase peptide synthesis and for preparing peptide mimetics. It is supplied with high purity and supplied under recommended storage conditions.

  • Fmoc-protected amino acid for peptide synthesis.
  • High purity suitable for research use (≈99.9%).
  • Molecular weight 391.46 g/mol and formula C24H25NO4.
  • Available as a 5 g package.
  • Store at room temperature as a solid; in solvent store at -80°C or -20°C per guidelines.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.