Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-L-photo-leucine | 1360651-24-6 | MFCD31380715 | 99.7% | 365.38 g/mol | C20H19N3O4 | 10 MG
SDP

Catalog No. 5000283061
Encompass_Preferred

Fmoc-L-photo-leucine is an Fmoc-protected leucine derivative that contains a photoreactive group for photo-crosslinking. It is supplied as a purified solid for incorporation into peptides and proteins by chemical synthesis to enable UV-activated covalent capture of interacting partners and to support structural and interaction studies.

  • Fmoc-protected amino acid for peptide synthesis.
  • Photo-crosslinking functionality for UV-activated covalent capture.
  • High purity (>99.7%) suitable for analytical and preparative use.
  • Soluble in DMSO for convenient stock preparation.
  • Molecular weight 365.38 g/mol for formulation calculations.

Catalog No. 50-002-83061 Supplier Medchemexpress LLC Supplier No. HYW14203010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-L-photo-leucine is an Fmoc-protected leucine derivative that contains a photoreactive group for photo-crosslinking. It is supplied as a purified solid for incorporation into peptides and proteins by chemical synthesis to enable UV-activated covalent capture of interacting partners and to support structural and interaction studies.

  • Fmoc-protected amino acid for peptide synthesis.
  • Photo-crosslinking functionality for UV-activated covalent capture.
  • High purity (>99.7%) suitable for analytical and preparative use.
  • Soluble in DMSO for convenient stock preparation.
  • Molecular weight 365.38 g/mol for formulation calculations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.