Learn More
Medchemexpress LLC Fmoc-L-photo-leucine | 1360651-24-6 | MFCD31380715 | 99.7% | 365.38 g·mol⁻¹ | C20H19N3O4 | 25 MG
SDP
Catalog No. 5000283059
Encompass_Preferred
Fmoc-L-photo-leucine is an Fmoc-protected, photo-reactive derivative of the amino acid leucine designed for incorporation into peptides by chemical synthesis. It enables site-specific photo-crosslinking and the introduction of a UV-activatable probe into peptides or proteins, supplied as a solid at high purity for research and analytical use.
  • Provides a photo-reactive leucine analogue for site-specific crosslinking.
  • Compatible with standard Fmoc solid-phase peptide synthesis protocols.
  • High purity suitable for analytical and preparative applications.
  • Solid form, white to off-white, for convenient storage and handling.
  • Soluble in DMSO for convenient dissolution and handling in synthesis.
Catalog No. 50-002-83059 Supplier Medchemexpress LLC Supplier No. HYW14203025MG
Fmoc-L-photo-leucine is an Fmoc-protected, photo-reactive derivative of the amino acid leucine designed for incorporation into peptides by chemical synthesis. It enables site-specific photo-crosslinking and the introduction of a UV-activatable probe into peptides or proteins, supplied as a solid at high purity for research and analytical use.
  • Provides a photo-reactive leucine analogue for site-specific crosslinking.
  • Compatible with standard Fmoc solid-phase peptide synthesis protocols.
  • High purity suitable for analytical and preparative applications.
  • Solid form, white to off-white, for convenient storage and handling.
  • Soluble in DMSO for convenient dissolution and handling in synthesis.