Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc-L-Photo-Leucine

Catalog No. 90739150MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-739-150MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-739-150MG Supplier Sigma Aldrich Supplier No. 90739150MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fmoc-L-Photo-Leucine is a diazirine-containing, Fmoc-protected leucine amino acid and multifunctional photo-crosslinker. Its incorporation into peptides or small-molecule probes and tools allows for photoaffinity labeling of cellular targets and protein-protein interactions upon UV light (≥360 nm) irradiation to form a covalent bond. This and other multifunctional probe building blocks will continue to accelerate drug discovery research for probing cellular mechanisms, target ID/validation, and understanding traditionally undruggable targets. An unprotected version is also available as 907278.Synthesis of a polymyxin derivative for photolabeling studies in the gram-negative bacterium Escherichia coliDeveloping diazirine-based chemical probes to identify histone modification 'readers' and 'erasers'Protein-Polymer Conjugation via Ligand Affinity and Photoactivation of Glutathione S-TransferaseFishing for Drug Targets: A Focus on Diazirine Photoaffinity Probe SynthesisPhoto-affinity labeling (PAL) in chemical proteomics: a handy tool to investigate protein-protein interactions (PPIs)

Specifications

CAS 945564-85-2
Molecular Formula C20H25BN4O2
Linear Formula C37H56N4O4
Quantity 10 mg
Synonym (1,3,5,5,7,9-hexamethyl-5H-4l4,5l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-10-yl)methyl 1H-imidazole-1-carboxylate; BODIPY Photocage
Molecular Weight (g/mol) 364.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.