Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc-L-Photo-Phe-Oh

Catalog No. 90729450MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-729-450MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-729-450MG Supplier Sigma Aldrich Supplier No. 90729450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fmoc-L-Photo-Phe-OH is a diazirine-containing, Fmoc-protected phenylalanine amino acid and multifunctional photo-crosslinker. Its incorporation into peptides or small-molecule probes and tools allows for photoaffinity labeling of cellular targets and protein-protein interactions upon UV light (≥360 nm) irradiation to form a covalent bond. This and other multifunctional probe building blocks will continue to accelerate drug discovery research for probing cellular mechanisms, target ID/validation, and understanding traditionally undruggable targets. An unprotected version is also available as 907340.Covalent modifier-type aggregation inhibitor of amyloid-beta based on a cyclo-KLVFF motifMode of Action of cGMP-dependent Protein Kinase-specific Inhibitors Probed by Photoaffinity Cross-linking Mass SpectrometryTrifluoromethyldiazirine: an effective photo-induced cross-linking probe for exploring amyloid formationSimple and Versatile Method for Tagging Phenyldiazirine PhotophoresFishing for Drug Targets: A Focus on Diazirine Photoaffinity Probe SynthesisPhoto-affinity labeling (PAL) in chemical proteomics: a handy tool to investigate protein-protein interactions (PPIs)

Specifications

CAS 1380314-24-8
Molecular Formula C40H63N7O10S
Linear Formula C78H65ClN2Pd
Quantity 50 mg
Synonym (2S,4R)-1-((S)-27-Azido-2-(tert-butyl)-4-oxo-6,9,12,15,18,21-hexaoxa-3-azaheptacosanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide; (S,R,S)-AHPC-2-2-2-2-2-2-6-N3; Crosslinker-E3 Ligase ligand conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader; VH032 conjugate
Molecular Weight (g/mol) 834.03 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.