Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-L-Ser((Ac)4-β-D-Glc)-OH | 118358-38-6 | MFCD03452820 | ≥98.0% | 657.62 g/mol | C32H35NO14 | 25 MG
SDP

Catalog No. 5000283834
Encompass_Preferred

Fmoc-L-Ser((Ac)4-β-D-Glc)-OH is an Fmoc-protected serine derivative bearing a peracetylated β-D-glucosyl substituent. It is provided as a high-purity solid reagent for research use only and is used as a protected amino-acid building block in peptide and glycopeptide synthesis.

  • High purity: ≥98.0%.
  • Molecular weight: 657.62 g/mol.
  • Chemical formula: C32H35NO14.
  • Suitable for solid-phase peptide synthesis and glycopeptide assembly.
  • White to off-white solid; stable under recommended storage.
  • CAS number 118358-38-6 for unambiguous identification.

Catalog No. 50-002-83834 Supplier Medchemexpress LLC Supplier No. HYW14564625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-L-Ser((Ac)4-β-D-Glc)-OH is an Fmoc-protected serine derivative bearing a peracetylated β-D-glucosyl substituent. It is provided as a high-purity solid reagent for research use only and is used as a protected amino-acid building block in peptide and glycopeptide synthesis.

  • High purity: ≥98.0%.
  • Molecular weight: 657.62 g/mol.
  • Chemical formula: C32H35NO14.
  • Suitable for solid-phase peptide synthesis and glycopeptide assembly.
  • White to off-white solid; stable under recommended storage.
  • CAS number 118358-38-6 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.