Learn More
BLD PHARMATECH CO LIMITED Fmoc-L-Ser(tBu)-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.2-0.8 mmol/g
Catalog No. NC3965227
Encompass_Preferred
Fmoc-L-Ser(tBu)-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.2-0.8 mmol/g
Catalog No. NC3965227 Supplier BLD PHARMATECH CO LIMITED Supplier No. BD3024685G
Fmoc-L-Ser(tBu)-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.2-0.8 mmol/g