Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-L-Ser tri-O-acetylGalNAc-, Advanced ChemTech

Catalog No. 01339729
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-339-729 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-339-729 Supplier Advanced Chem Tech Supplier No. GAA0171G
Only null left
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 1676104-71-4
Molecular Formula C32H36N2O13
Molecular Weight (g/mol) 656.64
InChI Key ORICVOOXZDVFIP-UKFUGAQESA-N
IUPAC Name (2S)-3-{[(2R,3R,4R,5R,6R)-4,5-bis(acetyloxy)-6-[(acetyloxy)methyl]-3-acetamidooxan-2-yl]oxy}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
SMILES CC(=O)N[C@H]1[C@H](OC[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)O[C@H](COC(C)=O)[C@H](OC(C)=O)[C@@H]1OC(C)=O

Specifications

Color White/Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-L-Ser tri-O-acetylGalNAc-
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.