Learn More
Selleck Chemical LLC Fmoc-Leu-d10 E8240-5MG
SDP
Catalog No. 5000040899
Encompass_Preferred
Fmoc-Leu-d10 is the deuterated form of Fmoc-Leu-OH Fmoc-Leu-OH is an intermediate used to protect leucine in solid phase peptide synthesis The peptide drugs synthesized by Fmoc-Leu-OH may have activities such as regulating insulin sensitivity
Catalog No. 50-000-40899 Supplier Selleck Chemical LLC Supplier No. E82405MG
Fmoc-Leu-d10 is the deuterated form of Fmoc-Leu-OH Fmoc-Leu-OH is an intermediate used to protect leucine in solid phase peptide synthesis The peptide drugs synthesized by Fmoc-Leu-OH may have activities such as regulating insulin sensitivity