Learn More
Medchemexpress LLC Fmoc-leucine-d10 | 1190594-22-9 | MFCD06658583 | 99.4% | 363.47 | C21H13D10NO4 | 10 MG
SDP
Catalog No. 5000211191
Encompass_Preferred
Fmoc-leucine-d10 is a deuterium-labeled, Fmoc-protected L-leucine supplied for use as an isotopically labeled building block in peptide synthesis and as an analytical standard. The material is provided in small laboratory quantities with supporting quality and safety documents available.
Catalog No. 50-002-11191 Supplier Medchemexpress LLC Supplier No. HY101064S310MG
Fmoc-leucine-d10 is a deuterium-labeled, Fmoc-protected L-leucine supplied for use as an isotopically labeled building block in peptide synthesis and as an analytical standard. The material is provided in small laboratory quantities with supporting quality and safety documents available.