Learn More
Medchemexpress LLC Fmoc-leucine-d3 | 538372-74-6 | 99.3% | 356.43 | C21H20D3NO4 | 25 MG
SDP
Catalog No. 5000213625
Encompass_Preferred
Fmoc-leucine-d3 is a deuterium-labeled N-Fmoc protected leucine used as a tracer and internal standard for quantitative analysis in analytical workflows. It is intended for research use in biochemical and pharmacological studies, including investigations of metabolic pathways and nuclear receptor modulation.
  • Deuterium-labeled amino acid suitable as tracer or internal standard.
  • High purity suitable for analytical applications.
  • Compatible with NMR, GC-MS, and LC-MS quantitation.
  • Available in small milligram pack sizes for research use.
  • Useful for metabolic and receptor modulation studies.
Catalog No. 50-002-13625 Supplier Medchemexpress LLC Supplier No. HY101064S225MG
Fmoc-leucine-d3 is a deuterium-labeled N-Fmoc protected leucine used as a tracer and internal standard for quantitative analysis in analytical workflows. It is intended for research use in biochemical and pharmacological studies, including investigations of metabolic pathways and nuclear receptor modulation.
  • Deuterium-labeled amino acid suitable as tracer or internal standard.
  • High purity suitable for analytical applications.
  • Compatible with NMR, GC-MS, and LC-MS quantitation.
  • Available in small milligram pack sizes for research use.
  • Useful for metabolic and receptor modulation studies.