Learn More
Apexbio Technology LLC Fmoc-Lys(2-Cl-Z)-OH 133970-31-7 5g
SDP
Catalog No. 5000092529
Encompass_Preferred
Fmoc-Lys 2-Cl-Z -OH (133970-31-7) is a lysine derivative designed for peptide synthesis It is used in peptide chemistry featuring an N-terminal 9-fluorenylmethyloxycarbonyl (Fmoc) group that protects the -amino group and a 2-chlorobenzyloxycarbonyl (2-Cl-Z) protection on the side-chain amino group This orthogonal protecting strategy enables selective deprotection steps supporting site-specific lysine modifications Based on these properties Fmoc-Lys 2-Cl-Z -OH is applicable in the construction of modified peptides for studies of structure activity relationships enzyme substrate interactions and peptide-based therapeutics in biomedical research
Catalog No. 50-000-92529 Supplier Apexbio Technology LLC Supplier No. A79185000
Fmoc-Lys 2-Cl-Z -OH (133970-31-7) is a lysine derivative designed for peptide synthesis It is used in peptide chemistry featuring an N-terminal 9-fluorenylmethyloxycarbonyl (Fmoc) group that protects the -amino group and a 2-chlorobenzyloxycarbonyl (2-Cl-Z) protection on the side-chain amino group This orthogonal protecting strategy enables selective deprotection steps supporting site-specific lysine modifications Based on these properties Fmoc-Lys 2-Cl-Z -OH is applicable in the construction of modified peptides for studies of structure activity relationships enzyme substrate interactions and peptide-based therapeutics in biomedical research